ENA189566470 Enamine

4-cyclopropaneamido-2-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD09044942
  • Cas Number : 926201-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43246
Availability details for
Quantity
  • Availability
  • Add to Cart