ENA189566472 Enamine

N1-(pyrimidin-2-yl)propane-1,3-diamine dihydrochloride

  • MDL number MFCD11858037
  • Cas Number : 1171176-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43248
Availability details for
Quantity
  • Availability
  • Add to Cart