ENA189566475 Enamine

3-(dimethylcarbamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09812754
  • Cas Number : 396733-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39737

Safety Information

signalword Danger
pcodes P260,P261,P264,P272,P280,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P333+P313,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-39737
pictograms GHS05,GHS07
hstatements H314,H317
Availability details for
Quantity
  • Availability
  • Add to Cart