ENA189566483 Enamine

2-{[(4-fluorophenyl)methyl]amino}propanoic acid

  • MDL number MFCD09836863
  • Cas Number : 1396962-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43270
Availability details for
Quantity
  • Availability
  • Add to Cart