ENA189566484 Enamine

tert-butyl N-(5-amino-2-fluorophenyl)carbamate

  • MDL number MFCD04114549
  • Cas Number : 535170-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43271
Availability details for
Quantity
  • Availability
  • Add to Cart