ENA189566493 Enamine

3-[(1,3-benzothiazol-2-yl)amino]propanoic acid

  • MDL number MFCD00520414
  • Cas Number : 104344-75-4
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43293
Availability details for
Quantity
  • Availability
  • Add to Cart