ENA189566494 Enamine

4-chloro-1-hydroxynaphthalene-2-carboxylic acid

  • MDL number MFCD00458680
  • Cas Number : 5409-15-4
  • Molecular Weight : 222.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43294
Availability details for
Quantity
  • Availability
  • Add to Cart