ENA189566498 Enamine

5-chloro-2-(piperidin-4-yl)-1,3-benzoxazole

  • MDL number MFCD10690319
  • Cas Number : 199292-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43299
Availability details for
Quantity
  • Availability
  • Add to Cart