ENA189566510 Enamine

1-[(pyridin-2-yl)methyl]piperidin-4-amine

  • MDL number MFCD08445804
  • Cas Number : 160357-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43312
Availability details for
Quantity
  • Availability
  • Add to Cart