ENA189566514 Enamine

4-[(1-methyl-1H-imidazol-2-yl)methoxy]benzaldehyde

  • MDL number MFCD05863357
  • Cas Number : 118001-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43321
Availability details for
Quantity
  • Availability
  • Add to Cart