ENA189566516 Enamine

3-fluoro-4-methyl-5-nitrobenzoic acid

  • MDL number MFCD11858054
  • Cas Number : 400-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43324
Availability details for
Quantity
  • Availability
  • Add to Cart