ENA189566518 Enamine

2-(2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-1-yl)acetic acid

  • MDL number MFCD11858055
  • Cas Number : 80310-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43327
Availability details for
Quantity
  • Availability
  • Add to Cart