ENA189566521 Enamine

6-(morpholin-4-yl)-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11858056
  • Cas Number : 1146290-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43331
Availability details for
Quantity
  • Availability
  • Add to Cart