ENA189566523 Enamine

3-[2-(1H-pyrazol-1-yl)ethoxy]aniline

  • MDL number MFCD11652470
  • Cas Number : 1152819-54-9
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43335
Availability details for
Quantity
  • Availability
  • Add to Cart