ENA189566526 Enamine

6-(2-methylpropoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09811820
  • Cas Number : 1016756-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43339
Availability details for
Quantity
  • Availability
  • Add to Cart