ENA189566528 Enamine

1-[(3-aminophenyl)methyl]piperidine-4-carboxamide

  • MDL number MFCD09816700
  • Cas Number : 1016764-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43341
Availability details for
Quantity
  • Availability
  • Add to Cart