ENA189566539 Enamine

propan-2-ylhydrazine hydrochloride

  • MDL number MFCD01746887
  • Cas Number : 16726-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43363

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-43363
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart