ENA189566549 Enamine

1-{2-[4-(4-fluorophenyl)piperazin-1-yl]pyridin-4-yl}methanamine

  • MDL number MFCD09806399
  • Cas Number : 1016712-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43378
Availability details for
Quantity
  • Availability
  • Add to Cart