ENA189566555 Enamine

1-difluoromethanesulfonyl-4-fluorobenzene

  • MDL number MFCD11857852
  • Cas Number : 153698-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41829
Availability details for
Quantity
  • Availability
  • Add to Cart