ENA189566560 Enamine

1-(5-fluoro-2-methoxyphenyl)ethan-1-amine

  • MDL number MFCD05215250
  • Cas Number : 35253-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43393
Availability details for
Quantity
  • Availability
  • Add to Cart