ENA189566566 Enamine

2-(2,4-difluorophenyl)oxirane

  • MDL number MFCD09741026
  • Cas Number : 111991-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43401
Availability details for
Quantity
  • Availability
  • Add to Cart