ENA189566567 Enamine

2-(2,5-difluorophenyl)oxirane

  • MDL number MFCD09736526
  • Cas Number : 954277-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43402
Availability details for
Quantity
  • Availability
  • Add to Cart