ENA189566571 Enamine

N'-hydroxy-4-methylpentanimidamide

  • Cas Number : 188719-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41895
Availability details for
Quantity
  • Availability
  • Add to Cart