ENA189566578 Enamine

5-fluoro-2-[3-(trifluoromethyl)phenyl]benzoic acid

  • MDL number MFCD09042206
  • Cas Number : 926200-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43432
Availability details for
Quantity
  • Availability
  • Add to Cart