ENA189566593 Enamine

octahydro-2H-1,4-benzoxazine

  • MDL number MFCD08273499
  • Cas Number : 52769-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43458
Availability details for
Quantity
  • Availability
  • Add to Cart