ENA189566601 Enamine

2-[3-(trifluoromethyl)phenoxy]ethan-1-amine hydrochloride

  • MDL number MFCD00793842
  • Cas Number : 104774-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43469
Availability details for
Quantity
  • Availability
  • Add to Cart