ENA189566606 Enamine

1-(prop-2-yn-1-yl)piperidine-2-carboxylic acid

  • MDL number MFCD08444860
  • Cas Number : 1032056-99-7
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43474
Availability details for
Quantity
  • Availability
  • Add to Cart