ENA189566611 Enamine

4-(1H-imidazol-1-yl)-2-(trifluoromethyl)aniline

  • MDL number MFCD09044735
  • Cas Number : 926202-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43485
Availability details for
Quantity
  • Availability
  • Add to Cart