ENA189566613 Enamine

ethyl 3-carbamoyl-6-(3-hydroxypropyl)pyrazolo[1,5-a]pyrimidine-7-carboxylate

  • MDL number MFCD11858088
  • Cas Number : 1146290-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43490
Availability details for
Quantity
  • Availability
  • Add to Cart