ENA189566614 Enamine

6-methylpiperidine-3-carboxamide

  • MDL number MFCD09927703
  • Cas Number : 89940-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43493
Availability details for
Quantity
  • Availability
  • Add to Cart