ENA189566622 Enamine

2-aminobutanenitrile hydrochloride

  • MDL number MFCD11858092
  • Cas Number : 93554-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-146
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43508
Availability details for
Quantity
  • Availability
  • Add to Cart