ENA189566634 Enamine

N'-hydroxy-2-(2-methylpiperidin-1-yl)ethanimidamide

  • Cas Number : 1016794-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43533
Availability details for
Quantity
  • Availability
  • Add to Cart