ENA189566637 Enamine

1-(1-aminoethylidene)amino 4-methyl benzene-1,4-dicarboxylate

  • Cas Number : 874452-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43538
Availability details for
Quantity
  • Availability
  • Add to Cart