ENA189566641 Enamine

1-[(2-chlorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09052273
  • Cas Number : 937608-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43544
Availability details for
Quantity
  • Availability
  • Add to Cart