ENA189566643 Enamine

1-(4-iodophenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD09040633
  • Cas Number : 20725-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43548
Availability details for
Quantity
  • Availability
  • Add to Cart