ENA189566646 Enamine

1-(5-methanesulfonyl-2-methylbenzenesulfonyl)piperazine hydrochloride

  • MDL number MFCD11857949
  • Cas Number : 1171728-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42739
Availability details for
Quantity
  • Availability
  • Add to Cart