ENA189566651 Enamine

5-methanesulfonyl-2-methylbenzene-1-sulfonamide

  • MDL number MFCD11212970
  • Cas Number : 1671-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42740
Availability details for
Quantity
  • Availability
  • Add to Cart