ENA189566652 Enamine

6-phenoxypyridine-3-carboxylic acid

  • MDL number MFCD02682068
  • Cas Number : 51362-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43572

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43572
Availability details for
Quantity
  • Availability
  • Add to Cart