ENA189566654 Enamine

1-(3,4-difluorophenyl)-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09043481
  • Cas Number : 926219-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43575
Availability details for
Quantity
  • Availability
  • Add to Cart