ENA189566663 Enamine

N-[(4-fluorophenyl)methyl]piperidine-4-carboxamide

  • MDL number MFCD08444679
  • Cas Number : 327048-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43593
Availability details for
Quantity
  • Availability
  • Add to Cart