ENA189566667 Enamine

N-(3-chloro-4-methylphenyl)piperazine-1-carboxamide hydrochloride

  • MDL number MFCD11858111
  • Cas Number : 1172020-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43599
Availability details for
Quantity
  • Availability
  • Add to Cart