ENA189566671 Enamine

1-nitro-4-trifluoromethanesulfonylbenzene

  • MDL number MFCD00185850
  • Cas Number : 432-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43603
Availability details for
Quantity
  • Availability
  • Add to Cart