ENA189566680 Enamine

3-(1H-imidazol-1-yl)-2-methylpropan-1-amine dihydrochloride

  • MDL number MFCD11858114
  • Cas Number : 1170402-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43627
Availability details for
Quantity
  • Availability
  • Add to Cart