ENA189566683 Enamine

1-phenyl-1H,4H,5H,6H,7H,8H-cyclohepta[c]pyrazole-3-carboxylic acid

  • MDL number MFCD09049895
  • Cas Number : 97393-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-168
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43653
Availability details for
Quantity
  • Availability
  • Add to Cart