ENA189566684 Enamine

1-cyclohexyl-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD09025531
  • Cas Number : 92905-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43654
Availability details for
Quantity
  • Availability
  • Add to Cart