ENA189566686 Enamine

3-ethoxy-4-methoxyaniline

  • MDL number MFCD09738742
  • Cas Number : 477742-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43656
Availability details for
Quantity
  • Availability
  • Add to Cart