ENA189566687 Enamine

5-chloro-2-(chloromethyl)-1-methyl-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD11858117
  • Cas Number : 1173070-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 301-302
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43658
Availability details for
Quantity
  • Availability
  • Add to Cart