ENA189566695 Enamine

1-phenyl-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD06799611
  • Cas Number : 43023-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43667
Availability details for
Quantity
  • Availability
  • Add to Cart