ENA189566700 Enamine

2-(quinolin-2-yl)acetonitrile

  • MDL number MFCD09924120
  • Cas Number : 14068-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Purity 91
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43680
Availability details for
Quantity
  • Availability
  • Add to Cart