ENA189566714 Enamine

5-chloro-2-methoxy-4-(1H-1,2,3,4-tetrazol-1-yl)benzoic acid

  • MDL number MFCD03419549
  • Cas Number : 926268-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-43722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43722
Availability details for
Quantity
  • Availability
  • Add to Cart